missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Golgi Glycoprotein 1/GLG1 Antibody (GLG1/7174R), CoraFluor™ 1, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-20874CL1
This item is not returnable.
View return policy
Description
Golgi Glycoprotein 1/GLG1 Monoclonal antibody specifically detects Golgi Glycoprotein 1/GLG1 in Human samples. It is validated for Immunohistochemistry (Paraffin),Immunohistochemistry (Frozen)
Specifications
| Golgi Glycoprotein 1/GLG1 | |
| Monoclonal | |
| CoraFluor 1 | |
| CFR1, CFR-1, cysteine-rich fibroblast growth factor receptor, DKFZp686L08213, E-selectin ligand 1, ESL1, ESL-1, FLJ23319, FLJ23967, FLJ32797, Golgi apparatus protein 1, golgi glycoprotein 1, golgi sialoglycoprotein MG-160, MG160, MG-160 | |
| Recombinant full-length human Golgi Glycoprotein 1/GLG1 protein | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| GLG1/7174R | |
| PBS | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| 2734 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction