missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Golgi Complex Antibody (GLG1/2829R), Allophycocyanin, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-08396APC
This item is not returnable.
View return policy
Description
Golgi Complex Monoclonal antibody specifically detects Golgi Complex in Human, Mouse (Negative), Rat (Negative) samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| Golgi Complex | |
| Monoclonal | |
| APC | |
| AI593353;AW537898;CFR;CFR1;CFR-1;ESL1;ESL-1;GLG1;MG160;MG-160;Selel | |
| Recombinant full-length human Golgi Complex protein (Uniprot: Q92896) | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| GLG1/2829R | |
| PBS | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Human, Mouse (Negative), Rat (Negative) | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction