missing translation for 'onlineSavingsMsg'
Learn More

GOLGA7 Antibody (2H8), Novus Biologicals™

Product Code. 18390829 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18390829 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18390829 Supplier Novus Biologicals Supplier No. H00051125M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

GOLGA7 Monoclonal antibody specifically detects GOLGA7 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation
TRUSTED_SUSTAINABILITY

Specifications

Antigen GOLGA7
Applications Western Blot, ELISA, Immunocytochemistry, Immunoprecipitation
Classification Monoclonal
Clone 2H8
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH12032
Gene Alias GCP16MGC21096, GOLGA3AP1, GOLGA7A, golgi autoantigen, golgin subfamily a, 7, Golgi complex-associated protein of 16 kDa, Golgi complex-associated protein of 16kDa, golgin A7, Golgin subfamily A member 7, HSPC041, MGC4876
Host Species Mouse
Immunogen GOLGA7 (AAH12032, 1 a.a. ~ 137 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MRPQQAPVSGKVFIQRDYSSGTRCQFQTKFPAELENRIDRQQFEETVRTLNNLYAEAEKLGGQSYLEGCLACLTAYTIFLCMETHYEKVLKKVSKYIQEQNEKIYAPQGLLLTDPIERGLRVIEITIYEDRGMSSGR
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51125
Target Species Human, Mouse
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.