missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GOLGA6L9 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15673335
Change view
Click to view available options
Quantity:
400 μL
Unit Size:
400µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15673335 400 μL 400µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15673335 Supplier Invitrogen™ Supplier No. PA549041

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

GOLGA6L9 Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen GOLGA6L9
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene GOLGA6L9
Gene Accession No. A6NEM1
Gene Alias GOLGA6L20; GOLGA6L9; golgi autoantigen, golgin subfamily a, 6-like 9; golgin A6 family-like 20; golgin A6 family-like 9; golgin subfamily A member 6-like protein 9; putative golgin subfamily A member 6-like protein 20
Gene Symbols GOLGA6L9
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 15-41 amino acids from the N-terminal region of human GOLGA6L9.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 440295
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.