missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GOLGA6B Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA562132
This item is not returnable.
View return policy
Description
Immunogen sequence: ALCAVSTQQQ EEDRSSSCRE AVLHRRLQQT IKERAL Highest antigen sequence identity to the following orthologs: Mouse - 36%, Rat - 33%.
This gene is found in a large, low copy repeat sequence or duplicon that is found in multiple copies, which are greater than 90% similar, on chromosome 15. Duplicons are associated with deletions, inversions and other chromosomal rearrangements that underlie genomic disease. This gene is a member of the golgin gene family, whose protein products localize to the Golgi apparatus. The majority of the related gene copies are thought to be transcribed pseudogenes. It is not known whether this gene is a pseudogene or if it encodes a golgin protein.
Specifications
| GOLGA6B | |
| Polyclonal | |
| Unconjugated | |
| GOLGA6B | |
| GOLGA; GOLGA6B; GOLGA6D; golgi autoantigen, golgin subfamily a, 6B; golgin A6 family member B; golgin A6 family, member B; golgin subfamily A member 6B; golgin-like protein; putative golgin subfamily A member 6B; Putative golgin subfamily A member 6D | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 55889 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| A6NDN3 | |
| GOLGA6B | |
| Recombinant protein corresponding to Human GOLGA6B. Recombinant protein control fragment (Product #RP-99520). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction