missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GOLGA6B Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-33588
This item is not returnable.
View return policy
Description
GOLGA6B Polyclonal specifically detects GOLGA6B in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| GOLGA6B | |
| Polyclonal | |
| Immunohistochemistry, Immunohistochemistry-Paraffin 1:20 - 1:50 | |
| A6NDN3 | |
| GOLGA6B | |
| This antibody was developed against a recombinant protein corresponding to amino acids: ALCAVSTQQQEEDRSSSCREAVLHRRLQQTIKERAL | |
| 0.1 mL | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| GOLGA, GOLGA6D, Golgi Autoantigen, Golgin Subfamily A, 6B, Golgin A6 Family, Member B, Golgin Subfamily A Member 6B, Golgin-Like Protein, Putative Golgin Subfamily A Member 6B, Putative Golgin Subfamily A Member 6D | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 55889 | |
| Human | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
For Research Use Only
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido