missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GOLGA4 Recombinant Rabbit Monoclonal Antibody (1I4T6)
GREENER_CHOICE

Rabbit Recombinant Monoclonal Antibody

Brand:  Invitrogen™ MA558088

 View more versions of this product

Product Code. 30282850

  • £363.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: MFKKLKQKIS EEQQQLQQAL APAQASSNSS TPTRMRSRTS SFTEQLDEGT PNRESGDTQS FAQKLQLRVP SVESLFRSPI KESLFRSSSK ESLVRTSSRE SLNRLDLDSS TASFDPPSDM DSEAEDLVGN SDSLNKEQLI QRLRRMERSL.

The Golgi apparatus, which participates in glycosylation and transport of proteins and lipids in the secretory pathway, consists of a series of stacked cisternae (flattened membrane sacs). Interactions between the Golgi and microtubules are thought to be important for the reorganization of the Golgi after it fragments during mitosis. This gene encodes one of the golgins, a family of proteins localized to the Golgi. This protein has been postulated to play a role in Rab6-regulated membrane-tethering events in the Golgi apparatus. Alternatively spliced transcript variants encoding different isoforms have been identified in this gene.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

GOLGA4
Recombinant Monoclonal
1.26 mg/mL
PBS with 0.05% BSA, 50% glycerol and 0.09% sodium azide; pH 7.3
Q13439, Q5U4E6, Q91VW5
GOLGA4
Recombinant fusion protein containing a sequence corresponding to amino acids 1-150 of human GOLGA4 (NP_002069.2).
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
ELISA, Immunohistochemistry (Paraffin)
1I4T6
Unconjugated
GOLGA4
256 kDa golgin; 72.1 protein; AI225887; AU019508; centrosome-related protein F46; CRPF46; EMBL:AAH85124.1, ECO:0000312; GCP2; GOLG; Golga4; golga4 {ECO:0000312; golgi autoantigen, golgin subfamily a, 4; golgin A4; Golgin subfamily A member 4; golgin subfamily A member 4 {ECO:0000250; golgin-240; golgin-245; MU-RMS-40.18; Olp-1; p230; protein 72.1; RGD:1591925}; tGolgin-1; trans-Golgi p230; UniProtKB:Q13439}
Rabbit
Protein A
RUO
2803, 501069, 54214
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.