Learn More
GOLGA2/GM130 Rabbit anti-Human, CoraLite 594, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech CL594-11308-100UL
Description
The GOLGA2/GM130 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human GOLGA2/GM130. This antibody recognizes human antigen. The GOLGA2/GM130 antibody has been validated for the following applications: FC (Intra), IF analysis. This antibody is coralite™594.
Specifications
| GOLGA2/GM130 | |
| Polyclonal | |
| Rabbit | |
| Primary | |
| Human |
| Immunofluorescence, Flow Cytometry (Intracellular Staining) | |
| CoraLite 594 | |
| 100 μL | |
| 2801 | |
| IgG |