Learn More
Specac™ Golden Gate™ Specular Reflectance Top Plate
Product Code. 15220098
Encompass_Preferred
Click to view available options
For Use With (Equipment):
Golden Gate ATR Accessory GS10500
Product Code. 15220098
Brand: Specac™ GS10514
This item requires a price update prior to being ordered.
Request a Price for this item here.
Micro Specular Reflectance Top-Plate including reference mirror for use with Golden Gate ATR Accessory GS10500. Can be used in all FTIR instruments.
Specifications
For Use With (Equipment) Golden Gate ATR Accessory GS10500