Learn More
Specac™ Golden Gate™ Mk II ATR View-Thru Anvil/Bridge Assembly
Click to view available options
Type:
Anvil/Bridge Assembly
Description
View-Thru Anvil/bridge assembly for use with the Golden Gate Single Reflection ATR Accessory GS10500.. The view-thru anvil allows a sample to be viewed through a 4x lens system with a built-in reflective illuminator.
Specifications
Specifications
| For Use With (Equipment) | Golden Gate Single Reflection ATR Accessory GS10500 |
| Type | Anvil/Bridge Assembly |