Learn More
Specac™ Golden Gate™ Mk II ATR Stainless Steel Flat Anvil
Product Code. 15250108
Encompass_Preferred
Click to view available options
For Use With (Equipment):
Golden Gate ATR Accessory GS10500
Product Code. 15250108
Brand: Specac™ GS10549
Stainless steel flat anvil used in the Golden Gate ATR Accessory GS10500. This is used for fibres or fine wires. It is not self-levelling, which can be an advantage with this type of sample.
Specifications
For Use With (Equipment) Golden Gate ATR Accessory GS10500