Learn More
Specac™ Golden Gate™ Low Temperature Diamond Mk ll ATR Top Plate
Product Code. 15260118
Encompass_Preferred
Change view
Click to view available options
For Use With (Equipment):
Golden Gate ATR Accessory GS10500
Product Code. For Use With (Equipment)
15260118 Golden Gate ATR Accessory GS10500
1 options
Product Code. 15260118 Supplier Specac™ Supplier No. GS10590
Estimated Shipment: 09-10-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Low Temperature Top-Plate option for the Golden Gate ATR Accessory GS10500. This can provide measurements down to near liquid nitrogen temperature. The systems operates from -150°C to 80°C. The system uses a thermally insulated copper and stainless steel dewar in conjunction with an integral heater. High thermal conductivity provides rapid cooling and temperature stabilisation.
Specifications
For Use With (Equipment) Golden Gate ATR Accessory GS10500
Description The systems operates from -150°C to +80°C.