Learn More
Specac™ Golden Gate™ High Temperature Diamond Mk ll ATR Top Plate
Description
High Temperature Diamond Top-Plate for use with the Golden Gate ATR Accessory. This Top-Plate can be heated to 300°C for reactions and processes that occur at high temperatures. Rapid and efficient heating can be achieved with a high degree of temperature control. The Top-Plate can be used with a computer controlled temperature controller with a digital readout to 1C.
Specifications
Specifications
| For Use With (Equipment) | Golden Gate ATR Accessory |