Learn More
Gold wire, 14kt, yellow, 1.63mm (0.064 in.) dia., Au 58.3% min, Thermo Scientific Chemicals
Brand: Thermo Scientific Alfa Aesar 040329.BS
275.34 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Code : NEW
Additional Details : CAS Number : 7440-57-5 Weight : 0.01000kg
Description
Gold wire is used in jewelry and as a plated coating on a wide variety of electrical and mechanical components. It is also acts as a heat reflecting coating for glass as well as a decorative medium.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Yellow | |
1.63mm (0.064 in.) dia. | |
10 cm | |
% Concentration Cu>Zn>Ag, Au ≤58.3% | |
Wire |
Gold wire (14kt) | |
MFCD00003436 | |
Soluble in aqua regia,nitric acid and hydrochloric acid. Insoluble in water. | |
≈28g/m |
RUO – Research Use Only