missing translation for 'onlineSavingsMsg'
Learn More

Gold wire, 14kt, yellow, 1.63mm (0.064in) dia, Au 58.3% min

Product Code. 11343898
Change view
Click to view available options
Quantity:
10 cm
25 cm
50 cm
Unit Size:
10cm
25cm
50cm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11343898 50 cm 50cm
11333898 10 cm 10cm
11353898 25 cm 25cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11343898 Supplier Thermo Scientific Alfa Aesar Supplier No. 040329.BW
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Gold wire is used in jewelry and as a plated coating on a wide variety of electrical and mechanical components. It is also acts as a heat reflecting coating for glass as well as a decorative medium.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Gold wire (14kt)
Color Yellow
Name Note 1.63mm (0.064 in.) dia.
Quantity 50 cm
Weight ≈28g/m
Form Wire
MDL Number MFCD00003436
Solubility Information Soluble in aqua regia,nitric acid and hydrochloric acid. Insoluble in water.
Concentration or Composition (by Analyte or Components) % Concentration Cu>Zn>Ag, Au ≤58.3%

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.