missing translation for 'onlineSavingsMsg'
Learn More

Gold wire, 1.0mm (0.04in) dia, Premion™, 99.9985% (metals basis)

Product Code. 11321538
Click to view available options
Quantity:
2 cm
10 cm
50 cm
Unit Size:
10cm
2cm
50cm
This item is not returnable. View return policy

Product Code. 11321538

Brand: Thermo Scientific Alfa Aesar 010965.BW

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7440-57-5 | Au | 196.97 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-57-5
Molecular Formula Au
Molecular Weight (g/mol) 196.97
MDL Number MFCD00003436
InChI Key PCHJSUWPFVWCPO-UHFFFAOYSA-N
Synonym colloidal, powder, flake, leaf, gold, colloidal, burnish, shell, ci pigment metal 3, magnesium purple, c.i. pigment metal 3
PubChem CID 23985
ChEBI CHEBI:30050
IUPAC Name gold
SMILES [Au]

Specifications

Quantity 50 cm
Odor Odorless
Weight ≈15.15g/m
Form Wire
Formula Weight 196.97
Percent Purity 99.9985%
Grade Premion
Assay (metals basis)
Chemical Name or Material Gold wire, 1.0mm (0.04 in.) dia.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.