missing translation for 'onlineSavingsMsg'
Learn More

Gold Tin eutectic foil, 0.1mm (0.004 in.) thick x 43.8mm (1.72in) wide, 99.99% (metals basis), Thermo Scientific Chemicals

Product Code. 11383388
Change view
Click to view available options
Quantity:
10 cm
50 cm
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11383388 10 cm Each
11373388 50 cm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11383388 Supplier Thermo Scientific Alfa Aesar Supplier No. 036310.BS
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Gold Tin eutectic foil is used in electronic manufacturing. It can be made into dimensions much smaller than soft solders, thereby it is utilized in applications which require the highest level of precision.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Gold Tin eutectic foil
Name Note 0.1mm (0.004 in.) thick x 43.8mm (1.72 in.) wide
Avg. Mol. Wt. or Mol. Wt. Range Au:Sn; 80:20 wt%
Quantity 10 cm
Weight ≈6.37g/10cm
Form Foil
Assay Percent Notes (metals basis)
MDL Number MFCD02091709
Solubility Information Insoluble in water.
Percent Purity 99.99%

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.