Learn More
Gold Tin eutectic foil, 0.1mm (0.004 in.) thick x 43.8mm (1.72in) wide, 99.99% (metals basis), Thermo Scientific Chemicals
Description
Gold Tin eutectic foil is used in electronic manufacturing. It can be made into dimensions much smaller than soft solders, thereby it is utilized in applications which require the highest level of precision.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Chemical Name or Material | Gold Tin eutectic foil |
| Name Note | 0.1mm (0.004 in.) thick x 43.8mm (1.72 in.) wide |
| Avg. Mol. Wt. or Mol. Wt. Range | Au:Sn; 80:20 wt% |
| Quantity | 50 cm |
| Weight | ≈6.37g/10cm |
| Form | Foil |
| Assay Percent Notes | (metals basis) |
| MDL Number | MFCD02091709 |
| Solubility Information | Insoluble in water. |
| Percent Purity | 99.99% |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.