missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gold slug, 3.175mm (0.125in) dia x 6.35mm (0.25in) length, Premion™, 99.99% (metals basis), Thermo Scientific Chemicals
Click to view available options
Quantity:
1 Pc.
25 Pc.
5 Pc.
Description
Gold slugs are used in thermal evaporation and electrodes (quartz oscillator, light-emitting diodes, and laser diodes).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | Golden |
Appearance | Gold colored slug |
Density | 19.32g/cm3 at 20°C |
Assay Percent Range | 99.99% (Metals basis) |
Quantity | 5 Pc. |
Solubility Information | Soluble in hot sulfuric acid and aqua regia. Insoluble in water and acid. |
Odor | Odorless |
Form | Slug |
Chemical Name or Material | Gold |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'