missing translation for 'onlineSavingsMsg'
Learn More

Gold slug, 3.175mm (0.125in) dia x 6.35mm (0.25in) length, Premion™, 99.99% (metals basis)

Product Code. 11410320
Change view
Click to view available options
Quantity:
1 Pc.
25 Pc.
5 Pc.
Unit Size:
Each
Pack of 25
Pack of 5
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11410320 1 Pc. Each
15444755 5 Pc. Pack of 5
11420320 25 Pc. Pack of 25
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11410320 Supplier Thermo Scientific Alfa Aesar Supplier No. 013394.KG
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Form: Approx.0.97g/slug | CAS: 7440-57-5 | Au | 196.97 g/mol

Gold slugs are used in thermal evaporation and electrodes (quartz oscillator, light-emitting diodes, and laser diodes).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-57-5
Molecular Formula Au
Molecular Weight (g/mol) 196.97
MDL Number MFCD00003436
InChI Key PCHJSUWPFVWCPO-UHFFFAOYSA-N
Synonym colloidal, powder, flake, leaf, gold, colloidal, burnish, shell, ci pigment metal 3, magnesium purple, c.i. pigment metal 3
PubChem CID 23985
ChEBI CHEBI:30050
IUPAC Name gold
SMILES [Au]

Specifications

Color Golden
Appearance Gold colored slug
Quantity 1 Pc.
Odor Odorless
Assay Percent Range 99.99% (Metals basis)
Form Slug
Solubility Information Soluble in hot sulfuric acid and aqua regia. Insoluble in water and acid.
Chemical Name or Material Gold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.