missing translation for 'onlineSavingsMsg'
Learn More

Gold Platinum Palladium powder, APS <5 micron, 99.9% (metal s basis), Thermo Scientific Chemicals

Product Code. 11443820
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11443820

Brand: Thermo Scientific Alfa Aesar 089726.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Gold Platinum Palladium powder
Avg. Mol. Wt. or Mol. Wt. Range Au:Pt:Pd; 40:40:20 wt%
Quantity 5 g
Particle Size APS <5 Micron
Form Powder
Assay Percent Notes (metals basis)
MDL Number MFCD00192403
UN Number UN3089
Percent Purity 99.9%

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.