missing translation for 'onlineSavingsMsg'
Learn More

Gold Palladium wire, 0.2mm (0.008in) dia, annealed, 99.9% (metals basis), Thermo Scientific Chemicals

Brand:  Thermo Scientific Alfa Aesar 041206.BU

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 11349577

  • £68.20 / 25cm
In Stock
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Gold Palladium wire is used as a microelectrode device, which exhibits excellent responses to hydrogen in concentrations as low as 0.5% by volume. It is also involved in the enhanced sensitivity for the detection of hydrogen.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Gold Palladium wire
Au:Pd; 60:40 wt%
MFCD00003436
Insoluble in water.
Wire
0.2mm (0.008 in.) dia., Annealed, Temper: softest 0.154g/ft
(metals basis)
25 cm
99.9%
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Special Offers

Special Offers

RUO – Research Use Only