Learn More
Gold nanoparticles, 15nm, supplied in 0.1mM PBS, reactant-free, 99%, OD1, 520nm absorption
Description
Ideal for sensitive applications requiring a minimum amount of contaminating reactants. Well suited for protein conjugate development by passive adsorption to the gold nanoparticle surface. Conjugates can be used in blotting, electron microscopy and immunohistochemistry applications
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Spécification
| Color | Red |
| Quantity | 25 mL |
| Particle Size | 15nm |
| Sensitivity | Light sensitive |
| Form | Liquid |
| Percent Purity | 99% |
| Chemical Name or Material | Gold nanoparticles, supplied in 0.1mM PBS, reactant-free, 99%, OD1, 520nm absorption |
RUO – Research Use Only
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.