missing translation for 'onlineSavingsMsg'
Learn More

Gold(III) oxide, Premion™, 99.99% (metals basis), Au 88.6% min

Product Code. p-7048940
Click to view available options
Quantity:
0.25 g
1 g
5 g
Unit Size:
0.25g
1g
5g
This item is not returnable. View return policy

Product Code. 11316538

Brand: Thermo Scientific Alfa Aesar 012161.01

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1303-58-8 | Au2O3 | 441.93 g/mol

It is used in aerospace and electrochemical applications such as fuel cells in which they exhibit ionic conductivity. It acts as strong reducing agent in redox reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1303-58-8
Molecular Formula Au2O3
Molecular Weight (g/mol) 441.93
MDL Number MFCD00014173
InChI Key DDYSHSNGZNCTKB-UHFFFAOYSA-N
Synonym digold trioxide, unii-lw5y686rup, gold iii oxide, gold 3+ ; oxygen 2-, lw5y686rup, gold trioxide, gold oxide, gold sesquioxide, gold oxide,, gold iii oxide, premion
PubChem CID 164805
IUPAC Name gold(3+);oxygen(2-)
SMILES [O-2].[O-2].[O-2].[Au+3].[Au+3]

Specifications

Melting Point 150°C (decomposition)
Color Brown
Physical Form Powder
Quantity 0.25 g
Merck Index 14,4524
Solubility Information Insoluble in water; Soluble in HCl,HNO3and in NaCN solution.
Formula Weight 441.93
Percent Purity 99.99%
Grade Premion
Concentration or Composition (by Analyte or Components) Au 88.6% min.
Odor Odorless
Assay (metals basis)
Chemical Name or Material Gold(III) oxide
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.