missing translation for 'onlineSavingsMsg'
Learn More

Gold(III) chloride trihydrate, Honeywell Fluka™

Product Code. p-7072297 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
25 g
Unit Size:
10g
1g
25g
5g
Product Code. Quantity unitSize
15657880 1 g 1g
15677880 5 g 5g
15647880 10 g 10g
15667880 25 g 25g
4 options
This item is not returnable. View return policy

Product Code. 15657880

Brand: Honeywell-Fluka G40221G

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

ACS Reagent,≥49.0% Au basis

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16961-25-4
Molecular Formula AuCl4H
Molecular Weight (g/mol) 339.77
MDL Number MFCD00149904
InChI Key VDLSFRRYNGEBEJ-UHFFFAOYSA-K
IUPAC Name trichlorogold hydrochloride
SMILES Cl.Cl[Au](Cl)Cl

Specifications

Quantity 1 g
Linear Formula HAuCl4 · 3H2O
UN Number UN3260
Formula Weight 393.83g/mol
Percent Purity ≥49% (Au basis)
Chemical Name or Material Gold(III) chloride trihydrate
compliance-icons
Product Identifier
  • Gold(III) chloride trihydrate
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.