missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gold(III) Chloride, TRC
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Specifications
Specifications
| Chemical Name or Material | Gold(III) Chloride |
| CAS | 13453-07-1 |
| Molecular Formula | Au Cl3 |
| Quantity | 250 mg |
| Synonym | Gold(III) chloride, Gold trichloride |
| InChI Formula | InChI=1S/Au.3ClH/h;3*1H/q+3;;;/p-3 |
| SMILES | Cl[Au](Cl)Cl |
| IUPAC Name | trichlorogold |
| Molecular Weight (g/mol) | 303.326 |
| Formula Weight | 301.873 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction