missing translation for 'onlineSavingsMsg'
Learn More

Gold(III) Chloride, TRC

Product Code. 30396842
Click to view available options
Quantity:
1 g
Unit Size:
1g
This item is not returnable. View return policy

Product Code. 30396842

Brand: Toronto Research Chemicals TRCG0778351G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Gold(III) Chloride
CAS 13453-07-1
Molecular Formula Au Cl3
Quantity 1 g
Synonym Gold(III) chloride, Gold trichloride
InChI Formula InChI=1S/Au.3ClH/h;3*1H/q+3;;;/p-3
SMILES Cl[Au](Cl)Cl
IUPAC Name trichlorogold
Molecular Weight (g/mol) 303.326
Formula Weight 301.873
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.