missing translation for 'onlineSavingsMsg'
Learn More

Gold(III) chloride, Au 64.4% min

Product Code. 11346538
Click to view available options
Quantity:
0.25 g
1 g
5 g
Unit Size:
0.25g
1g
5g
This item is not returnable. View return policy

Product Code. 11346538

Brand: Thermo Scientific Alfa Aesar 012163.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13453-07-1 | AuCl3

It is used in colloidal gold solutions, in photography and as a print toning agent(gold toning), starting solution to form other gold compounds and a precursor for preparation of ultra pure gold metal. It is used in electroplating and electroless plating as an anode in an electric cell. Gold(III) chloride acts as a gold catalyst and cell body stains for bright field and dark field microscopy.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13453-07-1
Molecular Formula AuCl3
MDL Number MFCD00014172

Specifications

Melting Point 180°C (sublimation)
Color Red
Physical Form Crystalline
Quantity 5 g
UN Number UN3260
Merck Index 14,4521
Solubility Information Soluble in water,ethanol,ether and slightly soluble in liquid ammonia.
Formula Weight 303.33
Odor Odorless
Sensitivity Hygroscopic
Density 3.90
Assay Au ≥64.4%
Chemical Name or Material Gold(III) chloride
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.