missing translation for 'onlineSavingsMsg'
Learn More

Gold(III) acetate, 99.9% (metals basis)

Product Code. 11394707
Change view
Click to view available options
Quantity:
0.5 g
1 g
5 g
Unit Size:
0.5g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11394707 1 g 1g
11314717 0.5 g 0.5g
11304717 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11394707 Supplier Thermo Scientific Alfa Aesar Supplier No. 039742.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 15804-32-7 | C6H9AuO6 | 374.099 g/mol

Used in solar energy technologies. Used in petrochemical cracking and automotive catalysts, water treatment, plating, textiles, research and in optic, laser, crystal and glass applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15804-32-7
Molecular Formula C6H9AuO6
Molecular Weight (g/mol) 374.099
MDL Number MFCD00210614
InChI Key OTCKNHQTLOBDDD-UHFFFAOYSA-K
Synonym gold 3+ triacetate, gold iii acetate, gold acetate, acmc-1c3i2, triacetic acid gold iii salt, acetic acid, gold 3+ salt 3:1
PubChem CID 15952561
IUPAC Name gold(3+);triacetate
SMILES CC(=O)[O-].CC(=O)[O-].CC(=O)[O-].[Au+3]

Specifications

Color Brown
Physical Form Powder
Quantity 1 g
Assay Percent Range 99.9% (Metal basis)
Solubility Information Slightly soluble in water.
Formula Weight 374.1
Appearance Brown powder
Chemical Name or Material Gold(III) acetate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.