missing translation for 'onlineSavingsMsg'
Learn More

Gold(I) potassium cyanide, Premion™, 99.96% (metals basis), Au 67.6% min

Product Code. 11337397
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11337397 5 g 5g
11317397 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11337397 Supplier Thermo Scientific Alfa Aesar Supplier No. 012552.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

CAS: 13967-50-5

Gold potassium cyanide is an important gold source used in electrolytic gold plating. The primary application of gold potassium cyanide is in the electroplating of metals. In addition, gold potassium cyanide is also used in the decorative plating of jewelry. It is useful for photoreduction of gold ions using the ZnO nanopowder. It is useful for the preparation of gold-gold junction electrode in the voltammetric glucose detection.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13967-50-5
MDL Number MFCD00011414
Synonym Potassium dicyanoaurate(I); Potassium gold cyanide

Specifications

Melting Point (decomposition)
Color White
Physical Form Crystalline Powder
Quantity 5 g
Linear Formula KAu(CN)2
UN Number UN1588
Beilstein 6235525
Merck Index 14,7628
Solubility Information Soluble in water and slightly soluble in alcohol.
Formula Weight 288.33
Percent Purity 99.96%
Grade Premion
Concentration or Composition (by Analyte or Components) Au 67.6% min.
Odor Odorless
Density 3.45 g/mL
Assay (metals basis)
Chemical Name or Material Gold(I) potassium cyanide
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Gold(I) potassium cyanide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 1
  • Acute toxicity Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H290-May be corrosive to metals.
  • H300-Fatal if swallowed.
  • H310-Fatal in contact with skin.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H330-Fatal if inhaled.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • EUH032-Contact with acids liberates very toxic gas.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P330-Rinse mouth.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Gold(I) potassium cyanide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.