missing translation for 'onlineSavingsMsg'
Learn More

Gold(I) chloride, 99.9% (metals basis)

Product Code. p-7058856
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11319548

Brand: Thermo Scientific Alfa Aesar 043365.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10294-29-8 | AuCl | 232.42 g/mol

Gold(I) chloride is used as a starting material for the preparation of Phosphine ligated gold(I) complexes, which is used as catalysts in synthetic chemistry. It is also used to prepare metallic gold and potassium auric bromide and potassium chloride.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10294-29-8
Molecular Formula AuCl
Molecular Weight (g/mol) 232.42
MDL Number MFCD00046175
InChI Key FDWREHZXQUYJFJ-UHFFFAOYSA-M
Synonym gold i chloride, gold monochloride, gold chloride, aurous chloride, gold chloride trihydride, acmc-1bvpa, gold monochloride, acid, gold i chloride trace metals basis, gold i chloride, saj special grade
PubChem CID 27366
ChEBI CHEBI:30078
IUPAC Name chlorogold
SMILES [Cl-].[Au+]

Tekniske data

Melting Point 289°C (decomposition)
Physical Form Powder
Quantity 1 g
UN Number UN3260
Merck Index 14,4515
Solubility Information Soluble in hydrochloric acid,hydrobromic acid and organic solvents. Slightly soluble in water.
Formula Weight 232.42
Percent Purity 99.9%
Odor Odorless
Sensitivity Moisture sensitive
Density 7.57 g/mL
Assay (metals basis)
Chemical Name or Material Gold (I) chloride
Vis mere Vis mindre

RUO – Research Use Only

Korrektion af produktindhold

Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.

Produkttitel

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.

Mange tak! Din feedback er blevet sendt. Hos Fisher Scientific arbejder vi altid på at forbedre vores indhold for dig. Vi sætter pris på din feedback.