missing translation for 'onlineSavingsMsg'
Learn More

Gold Germanium slug, 6.35mm (0.25 in.) dia. x 6.35mm (0.25 in.) length, Premion™, 99.9997% (metals basis), Thermo Scientific™

Brand:  Thermo Scientific Alfa Aesar 014172.KG

 View more versions of this product

Product Code. 15463495

  • £639.00 / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Gold Germanium slug
Au:Ge; 88:12 wt%
MFCD02091706
99.9997%
Odorless
Slug
6.35mm (0.25 in.) dia. x 6.35mm (0.25 in.) length
(metals basis)
1 Pc.
Premion™
≈2.9g/slug
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers