missing translation for 'onlineSavingsMsg'
Learn More

Gold Germanium slug, 3.175mm (0.125 in.) dia. x≈3.175mm (0.125 in.) length, Premion™, 99.998% (metals basis)

Product Code. p-7058668
Click to view available options
Quantity:
25 Pc.
5 Pc.
Unit Size:
Pack of 25
Pack of 5
This item is not returnable. View return policy

Product Code. 15413885

Brand: Thermo Scientific Alfa Aesar 042983.KJ

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Gold Germanium slug is used in scientific research and development activities.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Gold Germanium slug
Name Note 3.175mm (0.125 in.) dia. x≈3.175mm (0.125 in.) length
Avg. Mol. Wt. or Mol. Wt. Range Au:Ge; 88:12 wt%
Density 0.37 g/cm3
Quantity 5 Pc.
Odor Odorless
Weight ≈0.4g/slug
Form Slug
Assay Percent Notes (metals basis)
MDL Number MFCD02091706
Solubility Information Insoluble in water.
Percent Purity 99.998%
Grade Premion
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.