missing translation for 'onlineSavingsMsg'
Learn More

Gold gauze, 52 mesh woven from 0.102mm (0.004in) dia wire, 99.99% (metals basis)

Product Code. 11490341
Change view
Click to view available options
Quantity:
25 x 25 mm
50 x 50 mm
50 x 100 mm
100 x 100 mm
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11490341 50 x 50 mm Each
11345237 25 x 25 mm Each
15434835 50 x 100 mm Each
11335237 100 x 100 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11490341 Supplier Thermo Scientific Alfa Aesar Supplier No. 040930.FI
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Form: Wire cloth; Open area: 62.7%; Width of opening: 0.015 in. | CAS: 7440-57-5 | Au | 196.97 g/mol

Gold gauze, 52 mesh woven from 0.102mm (0.004in) dia wire, 99.99% (metals basis) is used as an electrode.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identifiants chimiques

CAS 7440-57-5
Molecular Formula Au
Molecular Weight (g/mol) 196.97
MDL Number MFCD00003436
InChI Key PCHJSUWPFVWCPO-UHFFFAOYSA-N
Synonym colloidal, powder, flake, leaf, gold, colloidal, burnish, shell, ci pigment metal 3, magnesium purple, c.i. pigment metal 3
PubChem CID 23985
ChEBI CHEBI:30050
IUPAC Name gold
SMILES [Au]

Spécification

Color Golden
Appearance Gold colored gauze
Quantity 50 x 50 mm
Mesh Size 52 Mesh
Odor Odorless
Assay Percent Range 99.99% (Metals basis)
Form Wire Cloth
Solubility Information Soluble in aqua regia.
Chemical Name or Material Gold

RUO – Research Use Only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.