missing translation for 'onlineSavingsMsg'
Learn More

Gold gauze, 100 mesh woven from 0.064mm (0.0025in) dia wire, 99.99% (metals basis), Thermo Scientific Chemicals

Product Code. 15421677
Click to view available options
Quantity:
25 x 25 mm
50 x 50 mm
50 x 100 mm
This item is not returnable. View return policy
This item is not returnable. View return policy

Form: Wire cloth 100 x 100mm Approx.4.9g; Open area: 56.3%; Width of opening: 0.0075 in.

Gold gauze, 100 mesh woven from 0.064mm (0.0025in) dia wire, 99.99% (metals basis) is used as an electrode.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-57-5
Molecular Formula Au
Molecular Weight (g/mol) 196.97
MDL Number MFCD00003436
InChI Key PCHJSUWPFVWCPO-UHFFFAOYSA-N
Synonym colloidal, powder, flake, leaf, gold, colloidal, burnish, shell, ci pigment metal 3, magnesium purple, c.i. pigment metal 3
PubChem CID 23985
ChEBI CHEBI:30050
IUPAC Name gold
SMILES [Au]

Specifications

Color Golden
Appearance Gold colored gauze
Assay Percent Range 99.99% (Metals basis)
Quantity 50 x 100 mm
Solubility Information Insoluble in water.
Mesh Size 100 Mesh
Odor Odorless
Form Wire Cloth
Chemical Name or Material Gold

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'