Learn More
Gold, AAS standard solution, Specpure™ Au 1000μg/mL
Description
Gold, AAS standard solution is used for the determination of gold by flame atomic absorption spectrometry (FAAS) and graphite furnace atomic absorption spectrometry (GFAAS).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Physical Form | Liquid |
| Concentration | Au in 20% HCl |
| UN Number | UN1789 |
| Solubility | Miscible with water |
| Quantity | 100 mL |
| Grade | Specpure™ |
| Chemical Name or Material | Gold, AAS standard solution, Au 1000μg/mL |
Safety and Handling
- Gold, AAS standard solution, Specpure, Au 1000µg/ml
- Warning
- Serious eye damage/eye irritation Category 2
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H290-May be corrosive to metals.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- MIXTURE LIST-Contains : Hydrochloric acid
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.