missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
Specifications
| Assay Range | 4-1000 pg/ml |
| Product Type | ELISA Development Kit |
| Sample Type | Cell Culture Supernatant, Plasma, Serum |
| Specificity | The kit contains a matched pair of monoclonal antibodies (mAbs) specific for native and recombinant bovine IL-5. The mAbs cross-react with IL-5 from goat. |
| For Use With (Application) | ELISA |
| Gene ID (Entrez) | 3567 |
| Gene Symbol | IL5 |
| Quantity | 1 Kit (for 6 Plates) |
| Research Discipline | Asthma, Biologically Active Proteins, Cell Cycle and Replication, Cytokine Research, Immunology |
| Storage Requirements | Storage of components varies. See protocol for specific instructions. |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?