Learn More
Goat IgG VisUCyte HRP Polymer, R&D Systems™
GREENER_CHOICE
Donkey Polyclonal Antibody has been used in 2 publications
Brand:  R&D Systems VC004-250
1734.00 GBP valid until 2025-12-16
BEST PRICE on Promo! Use promo code "24090" to get your promotional price.
 View more versions of this product
Product Code. 18749528
  • £2160.00 / 250mL
Description
IgG Polyclonal specifically detects IgG in Goat samples. It is validated for Immunohistochemistry.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
IgG
Polyclonal
Immunohistochemistry
Immunoglobulin G, ImmunoglobulinG
Goat IgG
250 mL (2 x 125 mL)
Secondary
Detects heavy ( gamma ) chains on goat IgG and light chains on all goat immunoglobulins, based on IEP. No cross-reactivity is observed with non-immunoglobulin goat serum proteins or serum proteins from human, mouse or rat, based on IEP. VisUCyte™ HRP Polymer provides high sensitivity with low background in IHC.
IgG
Immunohistochemistry
HRP Polymer
Supplied as a solution in PBS with preservative.
Donkey
Affinity Purified
RUO
3500
6 months from date of receipt, 2 to 8 degreesC as supplied.