missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat IgG VisUCyte HRP Polymer, R&D Systems™

Donkey Polyclonal Antibody has been used in 2 publications
Brand: R&D Systems VC004-125
918.00 GBP valid until 2025-12-16
BEST PRICE on Promo! Use promo code "24090" to get your promotional price.
This item is not returnable.
View return policy
Description
IgG Polyclonal specifically detects IgG in Goat samples. It is validated for Immunohistochemistry.
Specifications
| IgG | |
| Polyclonal | |
| Immunohistochemistry | |
| Immunoglobulin G, ImmunoglobulinG | |
| Goat IgG | |
| 125 mL | |
| Secondary | |
| Detects heavy ( gamma ) chains on goat IgG and light chains on all goat immunoglobulins, based on IEP. No cross-reactivity is observed with non-immunoglobulin goat serum proteins or serum proteins from human, mouse or rat, based on IEP. VisUCyte™ HRP Polymer provides high sensitivity with low background in IHC. | |
| IgG |
| Immunohistochemistry | |
| HRP Polymer | |
| Supplied as a solution in PBS with preservative. | |
| Donkey | |
| Affinity Purified | |
| RUO | |
| 3500 | |
| 6 months from date of receipt, 2 to 8 degreesC as supplied. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction