missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat F(ab')2 anti-Mouse IgG F(ab')2 Secondary Antibody, Rhodamine (Pre-adsorbed), Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NB120-5885
This item is not returnable.
View return policy
Description
IgG F(ab')2 Polyclonal antibody specifically detects IgG F(ab')2 in Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunofluorescence, ELISA.
Specifications
| IgG F(ab')2 | |
| Polyclonal | |
| Flow Cytometry 1:1:10 - 1:1000, Immunocytochemistry/Immunofluorescence 1:1000 - 1:5000, Fluorophore-linked immunosorbent assay 1:10000 - 1:50000 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| No reaction was observed against anti-Pepsin, anti-Goat IgG F(c), Mouse IgG F(c) or Bovine, Horse, Human, Rabbit, Rat and Sheep Serum Proteins. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
| Rhodamine | |
| immunoglobulin heavy constant gamma 1 (G1m marker) | |
| Mouse IgG F(ab)2 fragment | |
| 0.5 mg | |
| Secondary | |
| Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction