missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat F(ab')2 anti-Human IgG Fc Secondary Antibody, Novus Biologicals™
Goat Polyclonal Antibody
£107.00 - £188.00
Specifications
| Antigen | IgG Fc |
|---|---|
| Dilution | Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, ELISA 1:100-1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/ Immunofluorescence 1:10 - 1:500, Immunoassay, Immunomicroscopy 1:10 - 1:500, Chemiluminescence Immunoassay (CIA) (CIA) |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
IgG Fc Polyclonal antibody specifically detects IgG Fc in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoassay, Immunomicroscopy, Chemiluminescence ImmunoassaySpecifications
| IgG Fc | |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| Goat | |
| Adaptive Immunity, Immunology | |
| 10 mM Sodium Phosphate, 0.15 M Sodium Chloride, pH 7.2 | |
| Purified Human IgG, Fc fragment | |
| Secondary | |
| Store at 4°C. Do not freeze. |
| Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, ELISA 1:100-1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/ Immunofluorescence 1:10 - 1:500, Immunoassay, Immunomicroscopy 1:10 - 1:500, Chemiluminescence Immunoassay (CIA) (CIA) | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| O75015, O75015, O75015 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title