Learn More
ZNF423, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of ZNF423.
Brand: Abnova PAB18686.100ug
Description
The protein encoded by this gene is a nuclear protein that belongs to the family of Kruppel-like C2H2 zinc finger proteins. It functions as a DNA-binding transcription factor by using distinct zinc fingers in different signaling pathways. Thus, it is thought that this gene may have multiple roles in signal transduction during development. [provided by RefSeq
Sequence: EPECDQKTSRALEDRSpecifications
| ZNF423 | |
| Polyclonal | |
| Unconjugated | |
| ELISA (1:16000) The optimal working dilution should be determined by the end user. | |
| ZNF423 | |
| ZNF423 | |
| A synthetic peptide corresponding to amino acids near N-terminus of human ZNF423. | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
| ELISA | |
| 0.5 mg/mL | |
| Goat polyclonal antibody raised against synthetic peptide of ZNF423. | |
| In Tris saline, pH 7.3 (0.02% sodium azide, 0.5% BSA) | |
| Ebfaz/KIAA0760/MGC138520/MGC138522/OAZ/Roaz/ZFP423/Zfp104 | |
| Goat | |
| Antigen affinity purification | |
| RUO | |
| 23090 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only