missing translation for 'onlineSavingsMsg'
Learn More

ZBTB24, Goat, Polyclonal Antibody, Abnova™

Goat polyclonal antibody raised against synthetic peptide of ZBTB24.

Brand:  Abnova PAB27526.100ug

Product Code. 16087507

  • £432.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Sequence: PVEKDENYDPKTED
Specifications

Specifications

ZBTB24
Polyclonal
Unconjugated
ELISA (1:8000) Western Blot (1-3ug/ml) The optimal working dilution should be determined by the end user.
ZBTB24
ZBTB24
A synthetic peptide corresponding to amino acids 232-245 at internal region of human ZBTB24.
100 μg
Primary
Human
Western Blot
0.5 mg/mL
Goat polyclonal antibody raised against synthetic peptide of ZBTB24.
In Tris saline, pH 7.3 (0.02% sodium azide, 0.5% BSA)
BIF1/PATZ2/ZNF450
Goat
Affinity Purified
RUO
9841
Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only