Learn More
WDFY1, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of WDFY1.
Marca: Abnova PAB6612.100ug
Descrizione
The FYVE domain mediates the recruitment of proteins involved in membrane trafficking and cell signaling to phosphatidylinositol 3-phosphate (PtdIns(3)P)-containing membranes. This gene encodes a protein which contains a single FYVE domain and multiple WD40 repeats. This protein is localized to endosomes. [provided by RefSeq
Sequence: VVGCSLATGFSPHSpecifica
| WDFY1 | |
| Polyclonal | |
| Unconjugated | |
| ELISA (1:32000) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (4-6 ug/mL) The optimal working dilution should be determined by the end user. | |
| WDFY1 | |
| WDFY1 | |
| A synthetic peptide corresponding to C-terminus of human WDFY1. | |
| 100 μg | |
| Primary | |
| This antibody is expected to recognize both reported isoforms (as represented by NP_065881 and NP_848127) | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Immunohistochemistry (PFA fixed) | |
| 0.5 mg/mL | |
| Goat polyclonal antibody raised against synthetic peptide of WDFY1. | |
| In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
| FENS-1/WDF1/ZFYVE17 | |
| Goat | |
| Antigen affinity purification | |
| RUO | |
| 57590 | |
| Human | |
| Liquid |
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
For Research Use Only