Learn More
USH1C, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of USH1C.
Marca: Abnova PAB6589.100ug
Descrizione
This gene encodes a scaffold protein that functions in the assembly of Usher protein complexes. The protein contains PDZ domains, a coiled-coil region with a bipartite nuclear localization signal and a PEST degradation sequence. Defects in this gene are the cause of Usher syndrome type 1C and nonsyndromic sensorineural deafness autosomal recessive type 18. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq
Sequence: DRKVAREFRHKVD-CSpecifica
| USH1C | |
| Polyclonal | |
| Unconjugated | |
| ELISA (1:128000) Western Blot (0.1-0.3 ug/mL) Immunohistochemistry (10 ug/mL) The optimal working dilution should be determined by the end user. | |
| USH1C | |
| USH1C | |
| A synthetic peptide corresponding to human USH1C. | |
| 100 μg | |
| Primary | |
| This antibody is expected to recognize both reported isoforms (NP_005700.2 and NP_710142.1). | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Immunohistochemistry (PFA fixed), Western Blot | |
| 0.5 mg/mL | |
| Goat polyclonal antibody raised against synthetic peptide of USH1C. | |
| In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
| AIE-75/DFNB18/NY-CO-37/NY-CO-38/PDZ-45/PDZ-73/PDZ-73/NY-CO-38/PDZ73/ush1cpst | |
| Goat | |
| Antigen affinity purification | |
| RUO | |
| 10083 | |
| Human | |
| Liquid |
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
For Research Use Only