Learn More
UROD, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of UROD.
Brand: Abnova PAB27625.100ug
Description
This gene encodes the fifth enzyme of the heme biosynthetic pathway. This enzyme is responsible for catalyzing the conversion of uroporphyrinogen to coproporphyrinogen through the removal of four carboxymethyl side chains. Mutations and deficiency in this enzyme are known to cause familial porphyria cutanea tarda and hepatoerythropoetic porphyria. [provided by RefSeq
Sequence: KARECVGKTVTLQGNSpecifications
| UROD | |
| Polyclonal | |
| Unconjugated | |
| ELISA (1:128000) Western Blot (0.1-0.3ug/ml) The optimal working dilution should be determined by the end user. | |
| UROD | |
| UROD | |
| A synthetic peptide corresponding to internal region of human UROD. | |
| 100 μg | |
| Primary | |
| Human |
| Western Blot | |
| 0.5 mg/mL | |
| Goat polyclonal antibody raised against synthetic peptide of UROD. | |
| In Tris saline, pH 7.3 (0.02% sodium azide, 0.5% BSA) | |
| PCT | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 7389 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only