Learn More
Uricase, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against native Uricase (Bacillus).
Brand:  Abnova PAB28038.100ug
Product Code. 16203840
  • £752.00 / 100µg
Specifications
Uricase
Polyclonal
Goat polyclonal antibody raised against native Uricase (Bacillus).
Lyophilized from 20mM potassium phosphate buffer, 150mM NaCl, pH 7.2 (0.01% sodium azide)
ABC3735
Native purified Bacillus Uricase.
RUO
3202700
Lyophilized
ELISA, Immunoprecipitation, Western Blot
Unconjugated
ELISA (1:5000 - 1:20000) Immunoprecipitation (1:100) Western Blot (1:500-1:5000) The optimal working dilution should be determined by the end user.
ABC3735
Goat
100 μg
Primary
Store at 4°C on dry atmosphere. After reconstitution with 0.1mL of deionized water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.