missing translation for 'onlineSavingsMsg'
Learn More

TRAK2 (ALS2CR3), Goat, Polyclonal Antibody, Abnova™

Goat polyclonal antibody raised against synthetic peptide of ALS2CR3.

Brand:  Abnova PAB7496.100ug

Product Code. 16070568

  • £432.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Sequence: CTSSPKMGVLKED
Specifications

Specifications

TRAK2 (ALS2CR3)
Polyclonal
Unconjugated
ELISA (1:32000) The optimal working dilution should be determined by the end user.
TRAK2
TRAK2
A synthetic peptide corresponding to human ALS2CR3.
100 μg
Primary
Human
Liquid
ELISA
0.5 mg/mL
Goat polyclonal antibody raised against synthetic peptide of ALS2CR3.
In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide)
ALS2CR3/CALS-C/GRIF-1/GRIF1/KIAA0549/OIP98
Goat
Antigen affinity purification
RUO
66008
Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only