Learn More
TGIF2, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of TGIF2.
Brand: Abnova PAB6859.100ug
Description
The protein encoded by this gene is a DNA-binding homeobox protein and a transcriptional repressor. The encoded protein appears to repress transcription by recruiting histone deacetylases to TGF beta-responsive genes. This gene is amplified and overexpressed in some ovarian cancers, and mutations in this gene can cause holoprosencephaly. [provided by RefSeq
Sequence: C-EMELQKQQDPSpecifications
| TGIF2 | |
| Polyclonal | |
| Unconjugated | |
| ELISA (1:16000) Western Blot (0.3-1 ug/mL) The optimal working dilution should be determined by the end user. | |
| TGIF2 | |
| Goat | |
| Antigen affinity purification | |
| RUO | |
| 60436 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| 0.5 mg/mL | |
| Goat polyclonal antibody raised against synthetic peptide of TGIF2. | |
| In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
| TGIF2 | |
| A synthetic peptide corresponding to human TGIF2. | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only