missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TERF2IP, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of TERF2IP.
Brand: Abnova PAB19708.100ug
This item is not returnable.
View return policy
Description
Sequence: C-GAQNVARRIEFRKKSpecifications
| TERF2IP | |
| Polyclonal | |
| Goat polyclonal antibody raised against synthetic peptide of TERF2IP. | |
| In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
| DRIP5/RAP1 | |
| Goat | |
| Antigen affinity purification | |
| RUO | |
| 54386 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| Unconjugated | |
| ELISA (1:64000) Western Blot (0.1-0.3 ug/mL) The optimal working dilution should be determined by the end user. | |
| TERF2IP | |
| TERF2IP | |
| A synthetic peptide corresponding to C-terminus of human TERF2IP. | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction